The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163034.1 | NP_001156506.1 | 18 - 19 | 2953 | 93 |
NM_020761.2 | NP_065812.1 | 22 - 23 | 3427 | 93 | |
GenBank mRNA | AB037724.1 | - | 18 - 19 | 2109 | 93 |
AB082951.1 | - | 22 - 23 | 2622 | 93 | |
AY090663.1 | - | 22 - 23 | 2984 | 93 | |
BC136652.1 | - | 22 - 23 | 2817 | 93 | |
BC136654.1 | - | 22 - 23 | 2817 | 93 |