The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014296.2 | NP_055111.1 | 3 - 4 | 634 | 162 |
XM_011533555.2 | XP_011531857.1 | 3 - 4 | 485 | 162 | |
XM_011533556.2 | XP_011531858.1 | 4 - 5 | 717 | 162 | |
XM_011533557.2 | XP_011531859.1 | 3 - 4 | 607 | 162 | |
XM_017006043.1 | XP_016861532.1 | 4 - 5 | 717 | 162 | |
XM_017006044.1 | XP_016861533.1 | 3 - 4 | 607 | 162 | |
XM_017006045.1 | XP_016861534.1 | 4 - 5 | 673 | 162 | |
GenBank mRNA | AB028639.1 | - | 3 - 4 | 615 | 162 |
AK314253.1 | - | 3 - 4 | 596 | 162 | |
BC056202.1 | - | 3 - 4 | 612 | 162 |