The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003853.3 | NP_003844.1 | 1 - 2 | 152 | 117 |
XM_011512087.2 | XP_011510389.1 | 2 - 3 | 251 | 117 | |
GenBank mRNA | AF077346.1 | - | 1 - 2 | 147 | 116 |
AK300026.1 | - | 1 - 2 | 252 | 116 | |
DN996214.1 | - | 1 - 2 | 166 | 117 |