The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267727.1 | NP_001254656.1 | 1 - 2 | 72 | 100 |
XM_011524536.2 | XP_011522838.1 | 1 - 2 | 98 | 100 | |
XM_017024364.1 | XP_016879853.1 | 1 - 2 | 97 | 100 | |
XM_017024367.1 | XP_016879856.1 | 1 - 2 | 98 | 100 | |
XM_017024368.1 | XP_016879857.1 | 1 - 2 | 98 | 100 | |
GenBank mRNA | BG700864.1 | - | 1 - 2 | 77 | 100 |