Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VEZF1
Entrez Gene ID
7716
Gene Name
vascular endothelial zinc finger 1
Gene Aliases
DB1, ZNF161
Chromosome Location
Chr.17: 57971547 - 57988254 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007146.2 NP_009077.2 5 - 6 1288 64
XM_005257643.1 XP_005257700.1 5 - 6 1285 64
XM_005257644.4 XP_005257701.1 6 - 7 1441 64
XM_017025017.1 XP_016880506.1 6 - 7 1492 64
XM_017025018.1 XP_016880507.1 6 - 7 1344 64
GenBank mRNA AA195154.1 - 1 - 2 145 64
BX648862.1 - 5 - 6 581 64
D28118.1 - 5 - 6 1171 64

View Full Product Details