Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK10
Entrez Gene ID
5655
Gene Name
kallikrein related peptidase 10
Gene Aliases
NES1, PRSSL1
Chromosome Location
Chr.19: 51012744 - 51020175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145888.2 NP_665895.1 1 - 2 71 59
GenBank mRNA AF024605.1 - 1 - 2 71 59

View Full Product Details