Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AHSP
Entrez Gene ID
51327
Gene Name
alpha hemoglobin stabilizing protein
Gene Aliases
EDRF, ERAF
Chromosome Location
Chr.16: 31527852 - 31528804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016633.3 NP_057717.1 1 - 2 113 78
GenBank mRNA AF112995.1 - 1 - 2 144 78
AF147435.1 - 1 - 2 72 78
AF208865.1 - 1 - 2 69 78
AF364517.1 - 1 - 2 69 78
BC035842.1 - 1 - 2 54 78
BG940214.1 - 1 - 2 68 78

View Full Product Details