The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020452.3 | NP_065185.1 | 20 - 21 | 2233 | 146 |
XM_005245355.2 | XP_005245412.1 | 19 - 20 | 2380 | 146 | |
XM_005245356.3 | XP_005245413.1 | 20 - 21 | 2333 | 146 | |
XM_017001871.1 | XP_016857360.1 | 19 - 20 | 2383 | 146 | |
XM_017001872.1 | XP_016857361.1 | 20 - 21 | 2332 | 146 | |
GenBank mRNA | AB032963.1 | - | 10 - 11 | 1365 | 146 |
AK304806.1 | - | 2 - 3 | 181 | 146 | |
AY302537.1 | - | 20 - 21 | 2233 | 146 | |
BC007837.2 | - | 6 - 7 | 851 | 146 | |
BC041573.1 | - | 6 - 7 | 851 | 146 |