The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020452.3 | NP_065185.1 | 25 - 26 | 3129 | 67 |
XM_005245355.2 | XP_005245412.1 | 24 - 25 | 3276 | 67 | |
XM_005245356.3 | XP_005245413.1 | 25 - 26 | 3229 | 67 | |
XM_017001871.1 | XP_016857360.1 | 24 - 25 | 3279 | 67 | |
XM_017001872.1 | XP_016857361.1 | 25 - 26 | 3228 | 67 | |
GenBank mRNA | AB032963.1 | - | 15 - 16 | 2261 | 67 |
AK304806.1 | - | 7 - 8 | 1077 | 67 | |
AY302537.1 | - | 25 - 26 | 3129 | 67 | |
BC007837.2 | - | 11 - 12 | 1747 | 67 | |
BC030288.1 | - | 1 - 2 | 163 | 67 | |
BC041573.1 | - | 11 - 12 | 1747 | 67 | |
BC063858.1 | - | 1 - 2 | 229 | 67 |