The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289175.1 | NP_001276104.1 | 2 - 3 | 153 | 125 |
NM_001289176.1 | NP_001276105.1 | 2 - 3 | 153 | 125 | |
NM_022553.5 | NP_072047.4 | 2 - 3 | 472 | 125 | |
XM_017011177.1 | XP_016866666.1 | 2 - 3 | 407 | 125 | |
XM_017011178.1 | XP_016866667.1 | 2 - 3 | 125 | 125 | |
XM_017011179.1 | XP_016866668.1 | 2 - 3 | 407 | 125 | |
GenBank mRNA | AK001725.1 | - | 2 - 3 | 150 | 125 |
AK222867.1 | - | 2 - 3 | 198 | 125 | |
AK297935.1 | - | 2 - 3 | 128 | 125 | |
AK299563.1 | - | 3 - 4 | 308 | 125 | |
AK300308.1 | - | 2 - 3 | 384 | 125 | |
AK300430.1 | - | 2 - 3 | 170 | 125 | |
AK304080.1 | - | 2 - 3 | 371 | 125 | |
AL559461.3 | - | 2 - 3 | 158 | 125 | |
AX775737.1 | - | 2 - 3 | 316 | 125 | |
BC032108.1 | - | 2 - 3 | 334 | 125 | |
BC040114.1 | - | 2 - 3 | 353 | 125 | |
BU167150.1 | - | 2 - 3 | 392 | 125 | |
DC400338.1 | - | 2 - 3 | 431 | 125 |