The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077500.1 | NP_001070968.1 | 1 - 2 | 109 | 60 |
GenBank mRNA | BM830577.1 | - | 1 - 2 | 109 | 60 |