The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135564.1 | NP_001129036.1 | 11 - 12 | 1445 | 73 |
NM_004506.3 | NP_004497.1 | 12 - 13 | 1499 | 73 | |
GenBank mRNA | AK225593.1 | - | 11 - 12 | 1371 | 73 |
AK225594.1 | - | 11 - 12 | 1371 | 73 | |
AK294624.1 | - | 11 - 12 | 1335 | 73 | |
AK316377.1 | - | 11 - 12 | 1286 | 73 | |
BC112323.1 | - | 12 - 13 | 1402 | 73 | |
BC121050.1 | - | 11 - 12 | 1302 | 73 | |
BC121051.1 | - | 12 - 13 | 1356 | 73 | |
BC128420.1 | - | 11 - 12 | 1302 | 73 | |
M65217.1 | - | 12 - 13 | 1400 | 73 |