Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSF2
Entrez Gene ID
3298
Gene Name
heat shock transcription factor 2
Gene Aliases
HSF 2, HSTF 2
Chromosome Location
Chr.6: 122399551 - 122433119 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135564.1 NP_001129036.1 9 - 10 1259 106
NM_004506.3 NP_004497.1 9 - 10 1259 106
GenBank mRNA AK225593.1 - 9 - 10 1185 106
AK225594.1 - 9 - 10 1185 106
AK294624.1 - 9 - 10 1149 106
AK316377.1 - 9 - 10 1100 106
BC112323.1 - 9 - 10 1162 106
BC121050.1 - 9 - 10 1116 106
BC121051.1 - 9 - 10 1116 106
BC128420.1 - 9 - 10 1116 106
M65217.1 - 9 - 10 1160 106

View Full Product Details