Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMYA5
Entrez Gene ID
202333
Gene Name
cardiomyopathy associated 5
Gene Aliases
C5orf10, SPRYD2, TRIM76
Chromosome Location
Chr.5: 79689339 - 79800226 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153610.4 NP_705838.3 11 - 12 11860 122
XM_017009212.1 XP_016864701.1 11 - 12 11728 122
GenBank mRNA AF533705.1 - 10 - 11 122
AL359338.1 - 3 - 4 122
AL831966.1 - 10 - 11 3993 122
AL831968.2 - 6 - 7 787 122
AL831986.1 - 9 - 10 3516 122
AL832021.1 - 10 - 11 4234 122
AL834252.2 - 10 - 11 3577 122
BC111529.2 - 10 - 11 122
BC111530.2 - 10 - 11 3170 122
BX647628.1 - 10 - 11 3564 122
BX647646.1 - 10 - 11 3564 122

View Full Product Details