The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153610.4 | NP_705838.3 | 12 - 13 | 12033 | 107 |
XM_017009212.1 | XP_016864701.1 | 12 - 13 | 11901 | 107 | |
GenBank mRNA | AF533705.1 | - | 11 - 12 | 2627 | 107 |
AL359338.1 | - | 4 - 5 | 783 | 107 | |
AL831966.1 | - | 11 - 12 | 4166 | 107 | |
AL831968.2 | - | 7 - 8 | 960 | 107 | |
AL831986.1 | - | 10 - 11 | 3689 | 107 | |
AL832021.1 | - | 11 - 12 | 4407 | 107 | |
AL832368.1 | - | 11 - 12 | 3640 | 107 | |
AL834252.2 | - | 11 - 12 | 3750 | 107 | |
BC111529.2 | - | 11 - 12 | 3343 | 107 | |
BC111530.2 | - | 11 - 12 | 3343 | 107 | |
BX647628.1 | - | 11 - 12 | 3737 | 107 | |
BX647646.1 | - | 11 - 12 | 3737 | 107 |