Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMYA5
Entrez Gene ID
202333
Gene Name
cardiomyopathy associated 5
Gene Aliases
C5orf10, SPRYD2, TRIM76
Chromosome Location
Chr.5: 79689339 - 79800226 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153610.4 NP_705838.3 10 - 11 11763 84
XM_017009212.1 XP_016864701.1 10 - 11 11631 84
GenBank mRNA AF533705.1 - 9 - 10 2357 84
AL359338.1 - 2 - 3 513 84
AL831966.1 - 9 - 10 3896 84
AL831968.2 - 5 - 6 690 84
AL831986.1 - 8 - 9 3419 84
AL832021.1 - 9 - 10 4137 84
AL832368.1 - 9 - 10 3370 84
AL834252.2 - 9 - 10 3480 84
BC111529.2 - 9 - 10 3073 84
BC111530.2 - 9 - 10 3073 84
BX647628.1 - 9 - 10 3467 84
BX647646.1 - 9 - 10 3467 84

View Full Product Details