The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032251.5 | NP_115627.6 | 19 - 20 | 3402 | 145 |
XM_006718519.3 | XP_006718582.1 | 19 - 20 | 3414 | 145 | |
XM_011544960.2 | XP_011543262.1 | 18 - 19 | 3318 | 145 | |
GenBank mRNA | AK074126.1 | - | 12 - 13 | 3056 | 145 |
AK090436.1 | - | 18 - 19 | 3615 | 145 | |
BC040232.1 | - | 6 - 7 | 959 | 145 | |
BC140822.1 | - | 19 - 20 | 3391 | 145 | |
BC140824.1 | - | 19 - 20 | 3391 | 145 | |
BC141866.1 | - | 19 - 20 | 3381 | 145 |