The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313994.1 | NP_001300923.1 | 6 - 7 | 761 | 129 |
NM_145004.6 | NP_659441.3 | 7 - 8 | 841 | 129 | |
XM_006716298.2 | XP_006716361.1 | 7 - 8 | 635 | 129 | |
XM_006716299.1 | XP_006716362.1 | 7 - 8 | 635 | 129 | |
XM_011544433.1 | XP_011542735.1 | 7 - 8 | 635 | 129 | |
XM_011544434.1 | XP_011542736.1 | 7 - 8 | 635 | 129 | |
XM_017013184.1 | XP_016868673.1 | 7 - 8 | 635 | 129 | |
XM_017013185.1 | XP_016868674.1 | 6 - 7 | 641 | 129 | |
GenBank mRNA | AB209690.1 | - | 7 - 8 | 622 | 129 |
AK122608.1 | - | 7 - 8 | 635 | 129 | |
AK300148.1 | - | 6 - 7 | 719 | 129 | |
AK313864.1 | - | 7 - 8 | 662 | 129 | |
AY172952.1 | - | 7 - 8 | 763 | 129 | |
AY358739.1 | - | 7 - 8 | 721 | 129 | |
BC026085.2 | - | 7 - 8 | 625 | 129 | |
BC026169.2 | - | 7 - 8 | 652 | 129 | |
BC028702.2 | - | 7 - 8 | 635 | 129 | |
BC030014.2 | - | 7 - 8 | 628 | 129 | |
BC030698.2 | - | 7 - 8 | 643 | 129 | |
BC034975.2 | - | 7 - 8 | 636 | 129 |