The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018397.4 | NP_060867.2 | 6 - 7 | 1561 | 83 |
XM_006713250.3 | XP_006713313.1 | 6 - 7 | 1343 | 83 | |
XM_006713251.3 | XP_006713314.1 | 6 - 7 | 1520 | 83 | |
XM_011533938.2 | XP_011532240.1 | 6 - 7 | 1377 | 83 | |
XM_011533939.2 | XP_011532241.1 | 6 - 7 | 2216 | 83 | |
XM_017006797.1 | XP_016862286.1 | 6 - 7 | 1374 | 83 | |
XM_017006798.1 | XP_016862287.1 | 6 - 7 | 1366 | 83 | |
GenBank mRNA | AJ272267.1 | - | 4 - 5 | 785 | 83 |
AK297575.1 | - | 2 - 3 | 244 | 83 | |
BC034502.1 | - | 6 - 7 | 1319 | 83 |