The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005308.2 | NP_005299.1 | 14 - 15 | 1768 | 67 |
XM_005269707.1 | XP_005269764.1 | 13 - 14 | 1675 | 67 | |
XM_005269708.1 | XP_005269765.1 | 13 - 14 | 1658 | 67 | |
GenBank mRNA | AK298261.1 | - | 11 - 12 | 1255 | 67 |
AK301450.1 | - | 13 - 14 | 1459 | 67 | |
AK309761.1 | - | 14 - 15 | 1799 | 67 | |
AK313012.1 | - | 14 - 15 | 1615 | 67 | |
BC064506.1 | - | 14 - 15 | 1762 | 67 | |
BC114448.1 | - | 1 - 2 | 284 | 67 | |
L15388.1 | - | 14 - 15 | 1755 | 67 |