The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042494.1 | NP_001035959.1 | 11 - 12 | 1841 | 119 |
NM_001042495.1 | NP_001035960.1 | 11 - 12 | 1516 | 119 | |
NM_001042496.1 | NP_001035961.1 | 11 - 12 | 1907 | 119 | |
NM_001042497.1 | NP_001035962.1 | 9 - 10 | 1610 | 119 | |
NM_005135.2 | NP_005126.1 | 10 - 11 | 1480 | 119 | |
NM_133647.1 | NP_598408.1 | 10 - 11 | 1655 | 119 | |
XM_006720793.3 | XP_006720856.1 | 9 - 10 | 2390 | 119 | |
XM_011522269.2 | XP_011520571.1 | 10 - 11 | 2537 | 119 | |
GenBank mRNA | AF105366.1 | - | 10 - 11 | 1655 | 119 |
AF108831.1 | - | 10 - 11 | 1389 | 119 | |
AF116242.1 | - | 10 - 11 | 1491 | 119 | |
AF477977.1 | - | 11 - 12 | 1848 | 119 | |
AF531258.1 | - | 9 - 10 | 1610 | 119 | |
AF531259.1 | - | 11 - 12 | 1848 | 119 | |
AF531260.1 | - | 11 - 12 | 1486 | 119 | |
AK128133.1 | - | 9 - 10 | 1163 | 119 | |
AK292550.1 | - | 11 - 12 | 1594 | 119 | |
AK315283.1 | - | 10 - 11 | 1452 | 119 | |
AL117500.1 | - | 7 - 8 | 1083 | 119 | |
BC051744.1 | - | 11 - 12 | 1841 | 119 | |
BC070107.1 | - | 11 - 12 | 1511 | 119 | |
BC126241.1 | - | 10 - 11 | 1388 | 119 | |
BX648195.1 | - | 10 - 11 | 2597 | 119 | |
DQ138323.1 | - | 11 - 12 | 1906 | 119 |