The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042494.1 | NP_001035959.1 | 15 - 16 | 2284 | 81 |
NM_001042495.1 | NP_001035960.1 | 15 - 16 | 1959 | 81 | |
NM_001042496.1 | NP_001035961.1 | 15 - 16 | 2350 | 81 | |
NM_001042497.1 | NP_001035962.1 | 13 - 14 | 2053 | 81 | |
NM_005135.2 | NP_005126.1 | 14 - 15 | 1923 | 81 | |
NM_133647.1 | NP_598408.1 | 14 - 15 | 2098 | 81 | |
XM_006720793.3 | XP_006720856.1 | 13 - 14 | 2833 | 81 | |
XM_011522269.2 | XP_011520571.1 | 14 - 15 | 2980 | 81 | |
GenBank mRNA | AF105366.1 | - | 14 - 15 | 2098 | 81 |
AF108831.1 | - | 14 - 15 | 1832 | 81 | |
AF116242.1 | - | 14 - 15 | 1934 | 81 | |
AF477977.1 | - | 15 - 16 | 2291 | 81 | |
AF531258.1 | - | 13 - 14 | 2053 | 81 | |
AF531259.1 | - | 15 - 16 | 2291 | 81 | |
AF531260.1 | - | 15 - 16 | 1929 | 81 | |
AK128133.1 | - | 13 - 14 | 1606 | 81 | |
AK292550.1 | - | 15 - 16 | 2037 | 81 | |
AK315283.1 | - | 14 - 15 | 1895 | 81 | |
AL117500.1 | - | 11 - 12 | 1526 | 81 | |
BC051744.1 | - | 15 - 16 | 2287 | 81 | |
BC070107.1 | - | 15 - 16 | 1954 | 81 | |
BC126241.1 | - | 14 - 15 | 1831 | 81 | |
BC126243.1 | - | 14 - 15 | 1831 | 81 | |
BX648195.1 | - | 14 - 15 | 3040 | 81 | |
DQ138323.1 | - | 15 - 16 | 2349 | 81 |