Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC12A6
Entrez Gene ID
9990
Gene Name
solute carrier family 12 member 6
Gene Aliases
ACCPN, KCC3, KCC3A, KCC3B
Chromosome Location
Chr.15: 34229996 - 34338064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042494.1 NP_001035959.1 20 - 21 2979 55
NM_001042495.1 NP_001035960.1 20 - 21 2654 55
NM_001042496.1 NP_001035961.1 20 - 21 3045 55
NM_001042497.1 NP_001035962.1 18 - 19 2748 55
NM_005135.2 NP_005126.1 19 - 20 2618 55
NM_133647.1 NP_598408.1 19 - 20 2793 55
XM_006720793.3 XP_006720856.1 18 - 19 3528 55
GenBank mRNA AF105366.1 - 19 - 20 2793 55
AF108831.1 - 19 - 20 2527 55
AF116242.1 - 19 - 20 2629 55
AF477977.1 - 20 - 21 2986 55
AF531258.1 - 18 - 19 2748 55
AF531259.1 - 20 - 21 2986 55
AF531260.1 - 20 - 21 2624 55
AK128133.1 - 18 - 19 2301 55
AK292550.1 - 20 - 21 2732 55
AK315283.1 - 19 - 20 2590 55
AL117500.1 - 16 - 17 2221 55
BC051744.1 - 20 - 21 2982 55
BC070107.1 - 20 - 21 2649 55
BC126241.1 - 19 - 20 2526 55
BC126243.1 - 19 - 20 2526 55
DQ138323.1 - 20 - 21 3044 55

View Full Product Details