The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042494.1 | NP_001035959.1 | 5 - 6 | 895 | 66 |
NM_001042495.1 | NP_001035960.1 | 5 - 6 | 570 | 66 | |
NM_001042496.1 | NP_001035961.1 | 5 - 6 | 961 | 66 | |
NM_001042497.1 | NP_001035962.1 | 3 - 4 | 664 | 66 | |
NM_005135.2 | NP_005126.1 | 4 - 5 | 534 | 66 | |
NM_133647.1 | NP_598408.1 | 4 - 5 | 709 | 66 | |
XM_011522269.2 | XP_011520571.1 | 4 - 5 | 1591 | 66 | |
GenBank mRNA | AF105366.1 | - | 4 - 5 | 709 | 66 |
AF108831.1 | - | 4 - 5 | 443 | 66 | |
AF116242.1 | - | 4 - 5 | 545 | 66 | |
AF477977.1 | - | 5 - 6 | 902 | 66 | |
AF531258.1 | - | 3 - 4 | 664 | 66 | |
AF531259.1 | - | 5 - 6 | 902 | 66 | |
AF531260.1 | - | 5 - 6 | 540 | 66 | |
AK292550.1 | - | 5 - 6 | 648 | 66 | |
AK315283.1 | - | 4 - 5 | 506 | 66 | |
AL117500.1 | - | 1 - 2 | 137 | 66 | |
BC051744.1 | - | 5 - 6 | 895 | 66 | |
BC070107.1 | - | 5 - 6 | 565 | 66 | |
BC126241.1 | - | 4 - 5 | 442 | 66 | |
BC126243.1 | - | 4 - 5 | 442 | 66 | |
BX648195.1 | - | 4 - 5 | 1651 | 66 | |
DQ138323.1 | - | 5 - 6 | 960 | 66 |