The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042494.1 | NP_001035959.1 | 3 - 4 | 664 | 165 |
NM_001042495.1 | NP_001035960.1 | 3 - 4 | 339 | 165 | |
NM_001042496.1 | NP_001035961.1 | 3 - 4 | 730 | 165 | |
NM_133647.1 | NP_598408.1 | 2 - 3 | 478 | 165 | |
XM_006720793.3 | XP_006720856.1 | 2 - 3 | 1360 | 165 | |
XM_011522269.2 | XP_011520571.1 | 2 - 3 | 1360 | 165 | |
GenBank mRNA | AF105366.1 | - | 2 - 3 | 478 | 165 |
AF116242.1 | - | 2 - 3 | 314 | 165 | |
AF477977.1 | - | 3 - 4 | 671 | 165 | |
AF531259.1 | - | 3 - 4 | 671 | 165 | |
AF531260.1 | - | 3 - 4 | 309 | 165 | |
AK292550.1 | - | 3 - 4 | 417 | 165 | |
BC051744.1 | - | 3 - 4 | 664 | 165 | |
BC070107.1 | - | 3 - 4 | 334 | 165 | |
BX648195.1 | - | 2 - 3 | 1420 | 165 | |
DQ138323.1 | - | 3 - 4 | 729 | 165 |