The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013330.4 | NP_037462.1 | 7 - 8 | 1007 | 68 |
NM_197972.2 | NP_932076.1 | 7 - 8 | 976 | 68 | |
NR_104229.1 | 7 - 8 | 1007 | 68 | ||
XM_005245106.4 | XP_005245163.1 | 5 - 6 | 585 | 68 | |
XM_017001099.1 | XP_016856588.1 | 7 - 8 | 993 | 68 | |
XM_017001100.1 | XP_016856589.1 | 5 - 6 | 573 | 68 | |
GenBank mRNA | AF153191.2 | - | 7 - 8 | 846 | 68 |
AK094513.1 | - | 7 - 8 | 959 | 68 | |
AK290701.1 | - | 7 - 8 | 841 | 68 | |
AK301917.1 | - | 7 - 8 | 990 | 68 | |
BC006983.1 | - | 7 - 8 | 838 | 68 |