The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267701.1 | NP_001254630.1 | 2 - 3 | 518 | 166 |
GenBank mRNA | DA619574.1 | - | 2 - 3 | 343 | 166 |