Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAV3
Entrez Gene ID
859
Gene Name
caveolin 3
Gene Aliases
LGMD1C, LQT9, VIP-21, VIP21
Chromosome Location
Chr.3: 8733800 - 8746765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001234.4 NP_001225.1 2 - 3 543 62
GenBank mRNA Y14747.1 - 2 - 3 477 62

View Full Product Details