The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030755.4 | NP_110382.3 | 3 - 4 | 439 | 83 |
GenBank mRNA | AB048246.1 | - | 3 - 4 | 431 | 83 |
AK075395.1 | - | 3 - 4 | 346 | 83 | |
AK312905.1 | - | 3 - 4 | 317 | 83 | |
AL080080.1 | - | 3 - 4 | 337 | 83 | |
AY358640.1 | - | 3 - 4 | 326 | 83 | |
BC036460.2 | - | 3 - 4 | 389 | 83 | |
BC056874.1 | - | 3 - 4 | 328 | 83 | |
DA203669.1 | - | 3 - 4 | 439 | 83 |