The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024756.2 | NP_079032.2 | 2 - 3 | 422 | 62 |
XM_005270153.1 | XP_005270210.1 | 2 - 3 | 461 | 62 | |
XM_006717970.3 | XP_006718033.1 | 2 - 3 | 2401 | 62 | |
GenBank mRNA | AK023527.1 | - | 2 - 3 | 369 | 62 |
AK293822.1 | - | 2 - 3 | 369 | 62 | |
AK304727.1 | - | 2 - 3 | 279 | 62 | |
BC064415.1 | - | 2 - 3 | 377 | 62 | |
BC072456.1 | - | 2 - 3 | 318 | 62 | |
BC094744.1 | - | 2 - 3 | 369 | 62 | |
DC416005.1 | - | 2 - 3 | 423 | 62 |