Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC42BPB
Entrez Gene ID
9578
Gene Name
CDC42 binding protein kinase beta
Gene Aliases
MRCKB
Chromosome Location
Chr.14: 102932379 - 103057405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006035.3 NP_006026.3 16 - 17 2578 112
XM_005268227.1 XP_005268284.1 16 - 17 2408 112
XM_005268228.1 XP_005268285.1 16 - 17 2408 112
XM_005268229.1 XP_005268286.1 16 - 17 2408 112
XM_005268230.4 XP_005268287.1 16 - 17 2408 112
XM_011537387.1 XP_011535689.1 16 - 17 2408 112
GenBank mRNA AB032950.3 - 16 - 17 2569 112
AF128625.1 - 16 - 17 2635 112
BC047871.1 - 16 - 17 2471 112
BC155541.1 - 16 - 17 2581 112

View Full Product Details