Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NETO1
Entrez Gene ID
81832
Gene Name
neuropilin and tolloid like 1
Gene Aliases
BCTL1, BTCL1
Chromosome Location
Chr.18: 72742314 - 72867667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201465.1 NP_001188394.1 4 - 5 756 88
NM_138966.3 NP_620416.1 4 - 5 756 88
XM_005266774.4 XP_005266831.1 4 - 5 694 88
XM_017026017.1 XP_016881506.1 4 - 5 618 88
XM_017026018.1 XP_016881507.1 4 - 5 618 88
XM_017026019.1 XP_016881508.1 4 - 5 778 88
XM_017026020.1 XP_016881509.1 4 - 5 782 88
GenBank mRNA AF448838.1 - 4 - 5 762 88
AL834354.1 - 2 - 3 375 88

View Full Product Details