The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152892.2 | NP_690852.1 | 1 - 2 | 272 | 71 |
GenBank mRNA | AK290967.1 | - | 1 - 2 | 171 | 71 |
AK313771.1 | - | 1 - 2 | 155 | 71 | |
AL133057.1 | - | 1 - 2 | 222 | 71 | |
BC030547.1 | - | 1 - 2 | 212 | 71 | |
HY037356.1 | - | 1 - 2 | 272 | 71 |