Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FAM49B
- Entrez Gene ID
-
51571
- Gene Name
- family with sequence similarity 49 member B
- Gene Aliases
-
BM-009, L1
- Chromosome Location
-
Chr.8: 129839593 - 130016697 on Build GRCh38
View Full Product Details