The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138383.2 | NP_612392.1 | 3 - 4 | 462 | 79 |
XM_005256236.4 | XP_005256293.2 | 3 - 4 | 618 | 79 | |
XM_011523450.2 | XP_011521752.1 | 4 - 5 | 366 | 79 | |
XM_011523453.2 | XP_011521755.2 | 3 - 4 | 618 | 79 | |
XM_011523454.2 | XP_011521756.2 | 3 - 4 | 618 | 79 | |
XM_017023879.1 | XP_016879368.1 | 3 - 4 | 618 | 79 | |
XM_017023880.1 | XP_016879369.1 | 3 - 4 | 618 | 79 | |
XM_017023881.1 | XP_016879370.1 | 3 - 4 | 618 | 79 | |
XM_017023882.1 | XP_016879371.1 | 3 - 4 | 618 | 79 | |
XM_017023883.1 | XP_016879372.1 | 3 - 4 | 618 | 79 | |
XM_017023884.1 | XP_016879373.1 | 3 - 4 | 618 | 79 | |
GenBank mRNA | AB115770.1 | - | 3 - 4 | 202 | 79 |
BU859607.1 | - | 3 - 4 | 462 | 79 |