The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001924.2 | NP_001001924.1 | 12 - 13 | 3861 | 167 |
NM_001001925.2 | NP_001001925.1 | 11 - 12 | 3699 | 167 | |
NM_001001931.2 | NP_001001931.1 | 9 - 10 | 1448 | 167 | |
NM_001166393.1 | NP_001159865.1 | 9 - 10 | 1093 | 167 | |
NM_020749.4 | NP_065800.1 | 7 - 8 | 1245 | 167 | |
XM_005273577.3 | XP_005273634.1 | 12 - 13 | 3773 | 167 | |
XM_005273578.3 | XP_005273635.1 | 12 - 13 | 3861 | 167 | |
XM_005273579.3 | XP_005273636.1 | 12 - 13 | 3773 | 167 | |
XM_005273580.3 | XP_005273637.1 | 11 - 12 | 3611 | 167 | |
XM_005273581.3 | XP_005273638.1 | 11 - 12 | 3699 | 167 | |
XM_005273582.3 | XP_005273639.1 | 11 - 12 | 3611 | 167 | |
XM_011544595.2 | XP_011542897.1 | 12 - 13 | 3773 | 167 | |
XM_017013698.1 | XP_016869187.1 | 13 - 14 | 3779 | 167 | |
XM_017013699.1 | XP_016869188.1 | 13 - 14 | 4001 | 167 | |
XM_017013700.1 | XP_016869189.1 | 12 - 13 | 3660 | 167 | |
XM_017013701.1 | XP_016869190.1 | 13 - 14 | 3777 | 167 | |
XM_017013702.1 | XP_016869191.1 | 11 - 12 | 3432 | 167 | |
XM_017013704.1 | XP_016869193.1 | 6 - 7 | 767 | 167 | |
GenBank mRNA | AB033114.1 | - | 11 - 12 | 3313 | 167 |
AF121259.1 | - | 7 - 8 | 885 | 167 | |
AF293357.1 | - | 7 - 8 | 1158 | 167 | |
AK024357.1 | - | 6 - 7 | 771 | 167 | |
AK125188.1 | - | 9 - 10 | 1448 | 167 | |
AK289750.1 | - | 7 - 8 | 1244 | 167 | |
AK294860.1 | - | 9 - 10 | 1034 | 167 | |
AK304105.1 | - | 10 - 11 | 1330 | 167 | |
AK314692.1 | - | 7 - 8 | 1179 | 167 | |
AL096842.1 | - | 12 - 13 | 2653 | 167 | |
BC033842.1 | - | 4 - 5 | 337 | 167 | |
BC136320.1 | - | 12 - 13 | 3612 | 167 | |
BC142971.1 | - | 7 - 8 | 1178 | 167 | |
BX648879.1 | - | 5 - 6 | 1751 | 167 |