Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MGST1
Entrez Gene ID
4257
Gene Name
microsomal glutathione S-transferase 1
Gene Aliases
GST12, MGST, MGST-I
Chromosome Location
Chr.12: 16347142 - 16377189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267598.1 NP_001254527.1 3 - 4 306 91

View Full Product Details