Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- BAZ2B
- Entrez Gene ID
-
29994
- Gene Name
- bromodomain adjacent to zinc finger domain 2B
- Gene Aliases
-
WALp4
- Chromosome Location
-
Chr.2: 159315312 - 159712439 on Build GRCh38
View Full Product Details