Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
Chromosome Location
Chr.2: 159315312 - 159712439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289975.1 NP_001276904.1 34 - 35 6508 74
NM_013450.3 NP_038478.2 35 - 36 6616 74
XM_005246488.2 XP_005246545.2 36 - 37 6597 74
XM_005246489.4 XP_005246546.2 35 - 36 7700 74
XM_005246492.4 XP_005246549.2 35 - 36 7695 74
XM_006712461.3 XP_006712524.2 39 - 40 7171 74
XM_011511038.2 XP_011509340.1 36 - 37 6695 74
XM_011511039.1 XP_011509341.1 33 - 34 6318 74
XM_011511042.1 XP_011509344.1 34 - 35 6246 74
XM_011511043.1 XP_011509345.1 32 - 33 6216 74
XM_011511044.1 XP_011509346.1 33 - 34 6243 74
XM_011511045.1 XP_011509347.1 33 - 34 6156 74
XM_011511047.1 XP_011509349.1 35 - 36 6503 74
XM_011511048.2 XP_011509350.1 35 - 36 6568 74
XM_011511050.1 XP_011509352.1 33 - 34 6051 74
XM_011511051.1 XP_011509353.1 33 - 34 6132 74
XM_011511052.1 XP_011509354.1 32 - 33 5955 74
XM_011511056.1 XP_011509358.1 24 - 25 3971 74
XM_017003918.1 XP_016859407.1 39 - 40 7161 74
XM_017003919.1 XP_016859408.1 39 - 40 7155 74
XM_017003920.1 XP_016859409.1 41 - 42 7372 74
XM_017003921.1 XP_016859410.1 36 - 37 6695 74
XM_017003922.1 XP_016859411.1 36 - 37 6591 74
XM_017003923.1 XP_016859412.1 34 - 35 6345 74
XM_017003924.1 XP_016859413.1 33 - 34 6243 74
XM_017003925.1 XP_016859414.1 35 - 36 6599 74
XM_017003926.1 XP_016859415.1 35 - 36 6457 74
XM_017003927.1 XP_016859416.1 35 - 36 6497 74
XM_017003928.1 XP_016859417.1 35 - 36 6407 74
XM_017003929.1 XP_016859418.1 35 - 36 6384 74
XM_017003930.1 XP_016859419.1 35 - 36 6297 74
XM_017003931.1 XP_016859420.1 35 - 36 6395 74
XM_017003932.1 XP_016859421.1 32 - 33 6048 74
XM_017003933.1 XP_016859422.1 34 - 35 6274 74
XM_017003934.1 XP_016859423.1 34 - 35 6169 74
XM_017003935.1 XP_016859424.1 33 - 34 6107 74
XM_017003936.1 XP_016859425.1 33 - 34 6061 74
XM_017003937.1 XP_016859426.1 28 - 29 4768 74
GenBank mRNA AB032255.1 - 30 - 31 5989 74
AB040909.2 - 34 - 35 6476 74
AL080173.1 - 6 - 7 1056 74

View Full Product Details