The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012393.2 | NP_036525.1 | 6 - 7 | 820 | 67 |
XM_006721546.2 | XP_006721609.1 | 6 - 7 | 843 | 67 | |
XM_017024760.1 | XP_016880249.1 | 5 - 6 | 622 | 67 | |
GenBank mRNA | AB002359.1 | - | 6 - 7 | 779 | 67 |
AK292402.1 | - | 6 - 7 | 814 | 67 | |
AK292804.1 | - | 6 - 7 | 820 | 67 | |
AK295895.1 | - | 5 - 6 | 622 | 67 | |
BC146768.1 | - | 6 - 7 | 779 | 67 | |
BC167158.1 | - | 6 - 7 | 779 | 67 |