The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002062.4 | NP_002053.3 | 5 - 6 | 571 | 91 |
NR_136562.1 | 5 - 6 | 571 | 91 | ||
NR_136563.1 | 5 - 6 | 571 | 91 | ||
XM_017010750.1 | XP_016866239.1 | 4 - 5 | 1106 | 91 | |
XM_017010751.1 | XP_016866240.1 | 4 - 5 | 1109 | 91 | |
GenBank mRNA | BC112126.1 | - | 5 - 6 | 583 | 91 |
BC113493.1 | - | 5 - 6 | 571 | 91 | |
KR138540.1 | - | 5 - 6 | 537 | 91 | |
L23503.1 | - | 5 - 6 | 536 | 91 | |
U01104.1 | - | 5 - 6 | 553 | 91 | |
U01156.1 | - | 5 - 6 | 511 | 91 | |
U01157.1 | - | 5 - 6 | 511 | 91 | |
U10037.1 | - | 5 - 6 | 637 | 91 |