Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB12
Entrez Gene ID
221527
Gene Name
zinc finger and BTB domain containing 12
Gene Aliases
Bat9, C6orf46, D6S59E, G10, NG35
Chromosome Location
Chr.6: 31899194 - 31902331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011514383.2 XP_011512685.2 1 - 2 69 66

View Full Product Details