The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015335.4 | NP_056150.1 | 5 - 6 | 676 | 74 |
XM_011538080.2 | XP_011536382.1 | 5 - 6 | 709 | 74 | |
XM_011538081.2 | XP_011536383.1 | 5 - 6 | 709 | 74 | |
XM_011538082.2 | XP_011536384.1 | 5 - 6 | 879 | 74 | |
XM_017019090.1 | XP_016874579.1 | 5 - 6 | 709 | 74 | |
GenBank mRNA | AF515599.1 | - | 5 - 6 | 676 | 74 |
AK127706.1 | - | 2 - 3 | 210 | 74 | |
AY338463.1 | - | 5 - 6 | 828 | 74 | |
BC130422.1 | - | 5 - 6 | 672 | 74 | |
CR749332.1 | - | 2 - 3 | 200 | 74 |