Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VCAN
Entrez Gene ID
1462
Gene Name
versican
Gene Aliases
CSPG2, ERVR, GHAP, PG-M, WGN, WGN1
Chromosome Location
Chr.5: 83471674 - 83582303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126336.2 NP_001119808.1 1 - 2 353 116
NM_001164097.1 NP_001157569.1 1 - 2 353 116
NM_001164098.1 NP_001157570.1 1 - 2 353 116
NM_004385.4 NP_004376.2 1 - 2 353 116
GenBank mRNA AB209491.1 - 1 - 2 236 116
AK307642.1 - 1 - 2 254 116
BC050524.1 - 1 - 2 236 116
BP332680.1 - 1 - 2 316 116
D32039.1 - 1 - 2 102 116
DA361670.1 - 1 - 2 353 116
U16306.1 - 1 - 2 263 116
U26555.1 - 1 - 2 263 116
X15998.1 - 1 - 2 263 116

View Full Product Details