The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126336.2 | NP_001119808.1 | 2 - 3 | 431 | 144 |
NM_001164097.1 | NP_001157569.1 | 2 - 3 | 431 | 144 | |
NM_001164098.1 | NP_001157570.1 | 2 - 3 | 431 | 144 | |
NM_004385.4 | NP_004376.2 | 2 - 3 | 431 | 144 | |
GenBank mRNA | AB209491.1 | - | 2 - 3 | 314 | 144 |
AK307642.1 | - | 2 - 3 | 332 | 144 | |
BC050524.1 | - | 2 - 3 | 314 | 144 | |
BP332680.1 | - | 2 - 3 | 394 | 144 | |
D32039.1 | - | 2 - 3 | 180 | 144 | |
DA361670.1 | - | 2 - 3 | 431 | 144 | |
U16306.1 | - | 2 - 3 | 341 | 144 | |
U26555.1 | - | 2 - 3 | 341 | 144 | |
X15998.1 | - | 2 - 3 | 341 | 144 |