Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EBF3
Entrez Gene ID
253738
Gene Name
early B-cell factor 3
Gene Aliases
COE3, EBF-3, O/E-2, OE-2
Chromosome Location
Chr.10: 129835232 - 129964274 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005463.2 NP_001005463.1 9 - 10 938 104
XM_005252667.3 XP_005252724.1 9 - 10 1385 104
XM_005252668.3 XP_005252725.1 9 - 10 1412 104
XM_005252669.3 XP_005252726.1 9 - 10 1385 104
XM_006717739.3 XP_006717802.1 9 - 10 1412 104
XM_006717740.3 XP_006717803.1 9 - 10 1412 104
XM_006717741.3 XP_006717804.1 9 - 10 1412 104
XM_006717742.3 XP_006717805.1 9 - 10 1412 104
XM_006717743.3 XP_006717806.1 9 - 10 1412 104
XM_006717744.3 XP_006717807.1 9 - 10 1412 104
XM_011539575.2 XP_011537877.1 4 - 5 406 104
XM_017016027.1 XP_016871516.1 9 - 10 1412 104
GenBank mRNA AL832828.1 - 9 - 10 865 104
BC126130.1 - 9 - 10 929 104
BC130479.1 - 9 - 10 929 104

View Full Product Details