Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRELID2
Entrez Gene ID
153768
Gene Name
PRELI domain containing 2
Gene Aliases
-
Chromosome Location
Chr.5: 145587325 - 145835369 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138492.5 NP_612501.3 2 - 3 397 141
GenBank mRNA BC012033.1 - 2 - 3 364 141

View Full Product Details