Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MED13L
Entrez Gene ID
23389
Gene Name
mediator complex subunit 13 like
Gene Aliases
MRFACD, PROSIT240, THRAP2, TRAP240L
Chromosome Location
Chr.12: 115958576 - 116277219 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015335.4 NP_056150.1 12 - 13 2404 86
XM_011538080.2 XP_011536382.1 12 - 13 2437 86
XM_011538081.2 XP_011536383.1 12 - 13 2437 86
XM_011538082.2 XP_011536384.1 12 - 13 2607 86
XM_017019090.1 XP_016874579.1 12 - 13 2437 86
GenBank mRNA AB028948.2 - 6 - 7 1471 86
AF515599.1 - 12 - 13 2404 86
AK127706.1 - 9 - 10 1938 86
AL133033.1 - 6 - 7 1444 86
AY338463.1 - 12 - 13 2556 86
BC130422.1 - 12 - 13 2400 86
CR749332.1 - 9 - 10 1928 86

View Full Product Details