The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005188.3 | NP_005179.2 | 13 - 14 | 2297 | 62 |
XM_017018511.1 | XP_016874000.1 | 12 - 13 | 1732 | 62 | |
GenBank mRNA | BC132733.1 | - | 13 - 14 | 2226 | 62 |
BC136463.1 | - | 13 - 14 | 2226 | 62 | |
X57110.1 | - | 13 - 14 | 2303 | 62 |