Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM49B
Entrez Gene ID
51571
Gene Name
family with sequence similarity 49 member B
Gene Aliases
BM-009, L1
Chromosome Location
Chr.8: 129839593 - 130016697 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256763.1 NP_001243692.1 1 - 2 263 88
NR_046359.1 1 - 2 263 88
XM_011517113.2 XP_011515415.1 1 - 2 208 88
XM_011517117.2 XP_011515419.1 1 - 2 207 88
XM_011517124.2 XP_011515426.1 1 - 2 212 88
XM_017013548.1 XP_016869037.1 1 - 2 147 88
GenBank mRNA BC003599.1 - 1 - 2 42 88
BC017297.2 - 1 - 2 145 88
BX419336.2 - 1 - 2 263 88

View Full Product Details